To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Detection limit:~0.10 mg/dl
- Cat. no.:75817-140
- Includes:Kit contains: Standard, Reagents, and 96-Well Plate
- Supplier No.:700460-192
Spécifications
À propos de cet article
For the measurement of creatinine levels in plasma and serum.
- Measure creatinine levels in plasma and serum
- Assay 40 samples in duplicate
- Assay range: 0.0625-50 µM
- Measure creatinine levels down to 0.1 mg/dl
- Plate-based colorimetric measurement (490-500 nm)
Creatinine is commonly measured in urine and is a key benchmark for the normalization of a variety of urinary biomarkers. Serum creatinine levels, however, are a useful indicator of renal function. In addition, abnormal creatinine levels have been implicated in diabetes, cardiovascular, and circulatory diseases. Cayman’s Creatinine (serum) Assay is designed to measure creatinine levels in plasma and serum. The assay relies on the Jaffe’ reaction, wherein a yellow/orange color forms when the metabolite is treated with alkaline picrate. The rate of color development is directly proportional to the concentration of creatinine in the sample and is measured at an absorbance between 490-500 nm. The kinetic nature of the assay eliminates interference from extraneous serum contaminants, such as lipids and bilirubin.