ContactUsLinkComponent
S-Nitrosylated Protein Detection Kit (Biotin Switch), Cayman Chemical
Catalog # 75816-684
CAS Number:  
undefined
S-Nitrosylated Protein Detection Kit (Biotin Switch), Cayman Chemical
Catalog # 75816-684
Supplier Number:  10006518-1
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Cat. no.:
    75816-684
  • Includes:
    Kit contains: Buffers, Blocking Reagent, Labeling Reagent, Reducing Reagent, and Detection Reagents.
  • Supplier No.:
    10006518-1
Specifications
About this item
For detection of S-Nitrosylated proteins.
  • A modified ‘Biotin-switch’ assay
  • Direct visualization of S-NO proteins in whole cells or tissues
  • Colorimetric or fluorometric detection
Nitric oxide (NO) is produced by three distinct isoforms of nitric oxide synthase and functions as a key signaling molecule in physiology and pathophysiology. NO transduces its effects by reacting either directly with heme and non-heme centers of proteins or indirectly via further oxidation to various reactive nitrogen species (RNS). Cayman’s S-Nitrosylated Protein Detection Kit (Biotin Switch) employs a modification of the Jaffrey et al. 'Biotin-switch' method to allow for the direct visualization of S-NO proteins in whole cells or tissues, as well as by western blot analysis. Using this method, free SH groups are first blocked (an addition of 125.1 amu per residue) and any S-NO bonds present in the sample are then cleaved. Biotinylation of the newly formed SH groups (an addition of 523.6 amu per residue) provides the basis for visualization using streptavidin-based colorimetric or fluorescence detection.