Wheatstone Bridge
Wheatstone Bridge
Supplier:   Wards
Wheatstone Bridge
Supplier:  Wards
Certificates
Wheatstone Bridge
Wheatstone Bridge
2 Options Available Below
Product Details & Documents
Bridge has broad, heavily plated brass strips mounted on polished hardwood base.
  • A resistance wire negligible temperature coefficient is stretched over a meter ruler with millimeter graduations
Terminals, mounted on five plated brass strips, provide means for connecting resistors and a voltage source. These strips are separated by gaps that can be eliminated (as required) by using two further brass strips. A robust “jockey” (moveable contact) with lead is supplied for taking readings on the wire. A sprung clip provides convenient storage for the jockey.
Ordering information: Required but not included is a power supply and a galvanometer.
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    Availability
    Price
  • Wheatstone Bridge
    1.13×80×50 mm (two gap)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $145.05
    $145.05
    Each
       $145.05
    Supplier Number: PH1156A
  • Jockey Pen
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $25.30
    $25.30
    Each
       $25.30
    Supplier Number: PH1162