ContactUsLinkComponent
Prostaglandin D₂ (PGD₂) ELISA Kit, Cayman Chemical
undefined
Prostaglandin D₂ (PGD₂) ELISA Kit, Cayman Chemical
  • Assay Type:
    Competitive
  • Target protein:
    Prostaglandin D₂ (PGD₂)
  • Detection method:
    Colorimetric at 405 - 420 nm
  • Developmental time:
    90 - 120 min
  • Detection range:
    19.5 - 2500 pg/ml
  • Sensitivity:
    80% B/B0: 0.45 ng/ml
  • Incubation time:
    18 hr
Frequently Bought Together