Zymit® Pro Enzyme Cleaner, International Products Corporation
Zymit® Pro is a biodegradable, neutral-pH cleaner formulated with a unique blend of protease enzymes, surfactants, and builders that work together to remove tough protein-based soils.
- Typical pH of 7.5
- Free-rinsing; does not leave interfering residue
- Biodegradable and phosphate-free
- NSF-registered as a USDA-A1 cleaner
- Fast, effective soil removal
- ISO-Certified Quality System
The enzymes dissolve the soils, and the detergents help lift and wash them away
Registered with NSF as an A1 cleaner and can be validated in FDA processes. Validation methods are available upon request.
Target soils include food, gelatin, and biological materials such as blood, fat, and tissue. Zymit® Pro is effective for cleaning filter membranes, metals, ceramics, plastics, medical instruments and devices, food processing equipment, and other surfaces.
: Accessories not included
Frequently Bought Together