Polyethylene Plug Caps, Starplex®
: Starplex
Nonsterile polyethylene plug style caps are designed for use with 5mL and 10mL vials
All caps are bulk packaged in bags.
Nonsterile polyethylene plug style caps are designed for use with 5mL and 10mL vials
All caps are bulk packaged in bags.
Product Family Options
- DésignationCap ColorSterility
- Plug Style CapsDark BlueNon sterile-: V9222
- Plug Style CapsGreenNon sterile-: V9195
- Plug Style CapsLight BlueNon sterile-: V9199
- Plug Style CapsRedNon sterile-: V9200
- Plug Style CapsWhiteNon sterile-: V9220
- Plug Style CapsYellowNon sterile-: V9221
- : V9195
- : V9200
- Plug Style CapsLight BlueNon sterilePack of 1,000$25.00Case of 6000$150.00: V9199