Accessories for Eppendorf Shaking Incubators
Supplier: Eppendorf
Jump to:
À propos de cet article
18 Options Available Below
Product Details & Documents
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: S44I040002
- Supplier Number: S44I040003
- Supplier Number: S44I040004
- Supplier Number: S44I040005
- Supplier Number: S44I040006
- Supplier Number: S44I040010
- Angled test tube rack holder, for user-supplied test tube racks and microplatesDiscontinued-Supplier Number: TTR-210
- Angled test tube rack holder, for user-supplied test tube racks and microplatesLog in pour la disponibilitéEach$695.00Supplier Number: TTR-210
- Clamp bundle for Innova 42R, pre-selected clamps for the universal platformDiscontinued-Supplier Number: M1190-9921
- Clamp bundle for Innova 42R, pre-selected clamps for the universal platformLog in pour la disponibilitéEach$1,250.00Supplier Number: M1190-9921
- Culture drawer, (height adjustable) prevents plates from dryingDiscontinued-Supplier Number: M1335-0501
- Culture drawer, (height adjustable) prevents plates from dryingLog in pour la disponibilitéEach$1,700.00Supplier Number: M1335-0501
- Supplier Number: S44I042002
- Platform Assembly, 2 L, 18 x 30"Discontinued-Supplier Number: -
- Supplier Number: S44I041001
- Platform Dr., 2 L (18 x 30")Discontinued-Supplier Number: -
- Supplier Number: S44I041002
- Supplier Number: TTR-215
- Supplier Number: S44I041005
- Remote AlarmDiscontinued-Supplier Number: -
- Supplier Number: S44I041006
- Rod Kit for Utility Carrier (one additional rod)Discontinued-Supplier Number: M1001-9920
- Supplier Number: S44I041003
- Space bar for TTR-210Discontinued-Supplier Number: TTR-215
- Supplier Number: S44I042001
- Supplier Number: S44I040001