Eppendorf® Assay/Reader Microplates | Avantor
Eppendorf® Assay/Reader Microplates
   Options durables disponibles
Supplier:   Eppendorf
Eppendorf® Assay/Reader Microplates
Supplier:  Eppendorf
   Options durables disponibles
Certificates
Eppendorf® Assay/Reader Microplates
3 Options Available Below
Product Details & Documents
Eppendorf Assay/Reader plates are designed for diverse microplate assays, optimizing absorbance, fluorescence, and chemiluminescence. With options for both top- and bottom-reading plate readers, they ensure clear signals even with low-concentration samples.
  • Black Eppendorf Microplates offer an excellent signal-to-noise ratio – for clear signals even with low-concentration samples
  • White Eppendorf Microplates are optimized for highest sensitivity in the detection of luminescence signals by maximizing reflection
  • Solid black and white assay plates are made of polypropylene resulting in high resistance to chemicals, mechanical stress, and temperature extremes
  • All plates are optimized for minimal autofluorescence and autoluminescence
  • Lot-certified PCR clean purity grade
  • Outstanding dimensional accuracy and high well-to well homogeneity for perfect automation suitability
Perfect plate for your microplate assay protocol. Eppendorf assay plate range has the plate for your application, optimized for measuring absorbance in the visible range, fluorescence and chemiluminescence. The range includes both solid plates for top-reading plate readers and versatile clear and black/clear plates for use with both top- and bottom-reading plate readers.
Experience clear signals even with low-concentration samples. Black microplates are ideal for fluorescence detection, with an excellent signal-to-noise ratio, providing low detection limits in assays. White microplates maximize reflection for high-sensitivity detection of luminescence or weak fluorescence.
Specifications for Product Family
Spécifications
  • Catalog No:
  • CA89107-696
  • 75874-658
  • 75874-660
  • 75874-668
  • 75874-672
  • 89107-704
  • 89107-706
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
Spécifications
Product Family Options
96-well
Informations sur le produit
  • Bottom Style
    Color
    Plate Color
    Sterility
    Availability
    Price
  • U-bottom
    Black wells
    White
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 80
    Case of 80 - $656.00
    $656.00
    Case of 80
       $656.00
    Supplier Number: 951040102
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA89107-696
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    75874-658
    Environmentally Preferable
    Environmental certification
  • Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    75874-660
    Environmentally Preferable
    Environmental certification
  • Flat bottom
    White wells
    Gray
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 80
    Case of 80 - $491.30
    $491.30
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Each
    Each - $491.30
    $491.30
    Prix promotionnel pour les nouveaux clients
    Case of 80
       $491.30
    La quantité minimale de commande est de 1
    Each
       $491.30
    La quantité minimale de commande est de 1
    Supplier Number: 951040137
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    75874-668
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Flat bottom
    Black wells
    White
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 240
    Case of 240 - $488.75
    $488.75
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Each
    Each - $488.75
    $488.75
    Prix promotionnel pour les nouveaux clients
    Case of 240
       $488.75
    La quantité minimale de commande est de 1
    Each
       $488.75
    La quantité minimale de commande est de 1
    Supplier Number: 951040196
    Spécifications:
    More
    Specifications
    Cat. No.
    75874-672
    Environmentally Preferable
    Environmental certification
  • V-bottom
    Black wells
    White
    Non sterile
    Discontinued
    Supplier Number: 951040260
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89107-704
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • V-bottom
    White wells
    Gray
    Non sterile
    Discontinued
    Supplier Number: 951040308
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    89107-706
    Environmentally Preferable
    Environmental certification
    Product available on GSA Advantage®