Eppendorf® Protein LoBind Plates | Avantor
Eppendorf® Protein LoBind Plates
   Options durables disponibles
Supplier:   Eppendorf
Eppendorf® Protein LoBind Plates
Supplier:  Eppendorf
   Options durables disponibles
Certificates
Eppendorf® Protein LoBind Plates
8 Options Available Below
Product Details & Documents
Eppendorf Protein LoBind Plates are designed for use in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results.
  • Eppendorf LoBind material ensures excellent sample recovery for improved assay results
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: Free of human DNA, DNase, RNase and PCR inhibitors
  • High-contrast Unique OptiTrack® matrix: up to 30% faster sample identification and fewer pipetting errors
  • RecoverMax® well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
  • Raised well rims and a smooth surface ensure reliable sealing in plates
Protein sample recovery can be maximized with Eppendorf Protein LoBind Plates. Eppendorf LoBind plates are designed with a special, two-component polymer mix, creating a hydrophilic surface to ensure optimal sample recovery rates. These plates are specially developed for protein research or sensitive proteomic test methods, providing superior results by reducing sample binding to the plate surface.
Purity grade: PCR Clean
Material: polypropylene (PP)
Autoclavable: 121 °C, 20 min
Centrifugation stability: 6000×g
Well color: Clear
Specifications for Product Family
Spécifications
  • Catalog No:
  • CA89085-654
  • CA89085-656
  • CA89085-638
  • CA89085-640
  • 75874-682
  • CA89208-776
  • CA89085-670
  • CA89085-672
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
Spécifications
Product Family Options
96-well plates
Informations sur le produit
  • Désignation
    Well Volume
    Color
    Packaging
    Availability
    Price
  • Deepwell plate
    500 µl
    White
    40 plates (5 bags × 8 plates)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 40
    Case of 40 - $1,430.00
    $1,430.00
    Case of 40
       $1,430.00
    Supplier Number: 951032107
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA89085-654
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Deepwell plate
    500 µl
    White
    120 plates (10 bags × 12 plates)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 120
    Case of 120 - $3,850.00
    $3,850.00
    Case of 120
       $3,850.00
    La quantité minimale de commande est de 1
    Supplier Number: 951032506
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA89085-656
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Deepwell plate
    1000 µl
    White
    20 plates (5 bags × 4 plates)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 20
    Case of 20 - $546.00
    $546.00
    Case of 20
       $546.00
    Supplier Number: 951032905
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA89085-638
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Deepwell plate
    1000 µl
    White
    80 plates (10 bags × 8 plates)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 80
    Case of 80 - $1,930.00
    $1,930.00
    Case of 80
       $1,930.00
    Supplier Number: 951033308
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA89085-640
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Deepwell plate
    2000 µl
    White
    20 plates (5 bags × 4 plates)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 20
    Case of 20 - $687.00
    $687.00
    Case of 20
       $687.00
    Supplier Number: 0030504305
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA89208-776
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
384-well plates
Informations sur le produit
  • Désignation
    Well Volume
    Color
    Packaging
    Availability
    Price
  • Microplate
    140 µl
    White
    240 plates (10 bags × 24 plates)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 240
    Case of 240 - $4,830.00
    $4,830.00
    Log in pour la disponibilité
    Each
    Each - $4,830.00
    $4,830.00
    Case of 240
       $4,830.00
    La quantité minimale de commande est de 1
    Each
       $4,830.00
    La quantité minimale de commande est de 1
    Supplier Number: 951040601
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    75874-682
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Deepwell plate
    200 µl
    White
    40 plates (5 bags × 8 plates)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 40
    Case of 40 - $1,560.00
    $1,560.00
    Case of 40
       $1,560.00
    Supplier Number: 951031305
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA89085-670
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Deepwell plate
    200 µl
    White
    120 plates (10 bags × 12 plates)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 120
    Case of 120 - $4,170.00
    $4,170.00
    Case of 120
       $4,170.00
    Supplier Number: 951031704
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA89085-672
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact