To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Protein Assay Kit
- Size:200 assays
- Method:FL360/450 nm
- Sample:Biological
- Processing time:10 minutes
- Cat. no.:75878-192
- Supplier No.:QFPR-200
Specifications
About this item
For quantitative fluorimetric determination of total protein.
- Linear detection range of 0.05 - 200 μg/mL BSA.
- Homogeneous "mix-incubate-measure" type assay Can be readily automated to test thousands of samples per day
The protein or peptide is known as the "building blocks of life" and is one of the most important macromolecules in life science
BioAssay Systems’ QuantiFluo™ protein assay kit is based on an improved o-phthalaldehyde method. This reagent reacts with primary amines in protein or peptide and forms a blue fluorescent product, allowing detection of nanograms of proteins. The fluorescence intensity (λex/em = 360/450nm) is proportional to the protein concentration in the sample.
Protein determination is a very common practice. Simple, direct and automation-ready. Procedures for measuring protein or peptide concentration are very desirable