ContactUsLinkComponent
EnzyChrom™ Xanthine Oxidase Assay Kit, BioAssay Systems
Catalog # 75878-180
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
EnzyChrom™ Xanthine Oxidase Assay Kit, BioAssay Systems
Catalog # 75878-180
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  EXOX-100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Xanthine Oxidase Assay Kit
  • Size:
    100 assays
  • Method:
    OD570 nm, FL530/585 nm
  • Sample:
    Cell lysate, serum, and other biological samples.
  • Processing time:
    20 minutes
  • Cat. no.:
    75878-180
  • Supplier No.:
    EXOX-100
Specifications
About this item
For quantitative determination of xanthine oxidase enzyme activity.
  • Linear detection range 0.03 - 25 U/L xanthine oxidase for colorimetric assays and 0.01 -2.5 U/L for fluorimetric assays
  • Convenient room temperature and high-throughput assay
Xanthine oxidase catalyzes the oxidation of xanthine to uric acid
BioAssay Systems’ xanthine oxidase assay kit uses a single Working Reagent that combines the xanthine oxidase reaction and color reaction in one step. The change in color intensity of the reaction product at 570 nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to xanthine oxidase activity in the sample.
In addition, xanthine oxidase can catalyze the oxidation of hypoxanthine to xanthine, act on certain purines and aldehydes, and in certain cases produce the superoxide ion. Clinically, xanthine oxidase activity in blood can act as a marker for influenza, liver damage, and possibly cardiovascular health. Simple, direct and high-throughput assays for measuring xanthine oxidase activity find wide applications in research and drug discovery.