À propos de cet article
Perform sample transfers between virtually any tube, rack, microplate or horizontal gel box quickly and efficiently with adjustable tip spacing.
- Innovative ClipTip interlocking tip interface technology
- Designed for optimum comfort and ease of use
- Two operating options
- Two-year manufacturer's warranty with convenient web registration
Perform sample transfers between virtually any tube, rack, microplate or horizontal gel box quickly and efficiently with the Thermo Scientific™ E1-ClipTip™ equalizer pipette.
Adjustable tip spacing allows you to set the distance between tips by simply sliding the scale to expand or contract to the desired labware format. Feel the difference. Advanced connectivity enables access to Thermo Fisher Scientific cloud-based pipetting apps. Tip is locked and sealed in place until ejected, so there are no dropped tips that waste valuable sample. Complete seal to secure the sample volume in each tip for enhanced accuracy and precision. Tips are attached and ejected with a light touch. This eliminates the frustration of banging tips on the pipette.
Customers can utilize my pipette creator, a new web-based pipetting app powered by Thermo Fisher cloud, to revolutionize the way you pipette with a new era of technology. This enables efficient, centralized programming and sharing of protocols between E1-ClipTip pipettes and colleagues. The app offers pre-programmed protocols, to download and share, for many of your favorite reagent kits and pipetting protocols. Connect E1-ClipTip pipette to the app via a Bluetooth or USB connection. Accelerate collaboration and reproducibility, all from the comfort of your computer.
E1-ClipTip is designed for optimum comfort and ease of use. The electronic tip ejection and index finger pipetting action let your thumb relax during pipetting. Step-based Matrix or Presets with settings for your most common applications are two operating options available. The Programs function allows you to store 30 your most common Matrix or Presets protocols with a specific name. Five different user profiles offer customized settings for multiple users and protocols. The MyPipette™ function allows you to customize your main menu view by organizing the icons in your preferred order or creating shortcuts for your favorite programs and pipetting functions. You can also protect important settings from unintentional editing with a password. Service and calibration trackers remind you when the pipette needs to be sent for service. Pipettors run on a Li-ion battery for long lasting performance and fast recharging. Recharging can be done with a charging stand or a plug.
Available formats include Microcentrifuge tubes to/from 24,-48,-96-and 384- well microplates; Test tubes to/from deepwell blocks; Microplates and tubes to agarose gels. Save up to 90% in time and repetitions compared to use of single channel manual pipettes.