Static Dissipative Anti-Fatigue Matting, Wearwell®
Fournisseur: Wearwell
Certificats
À propos de cet article
The static dissipative mat has a homogeneous carbon layer assuring speedy dissipation of static.
- Specially formulated homogeneous carbon layer ensures rapid dissipation of static electricity
- Nitricell sponge base delivers 2X the resilience and 3X the life of typical sponges
- Lay-flat design and up to 20% more standing area than look-alikes
- Grounding snap is attached to the corner of each mat
This static dissipative anti-fatigue mat is an ideal ergonomic standing solution for manufacturing, pharmaceutical, and commercial areas where static electricity can pose a problem. The homogeneous carbon layer, which assures the speedy dissipation of static, is a standout feature. Additionally, the exclusive Nitricell sponge base lasts for years while providing unparalleled ergonomic benefits.
Certifications: ASTM F1677. Dry: COF = 0.62. ESD Rating: EOS/ESD S7.1: RTT= 1 x 10⁶ to 1 x 10¹⁰.
Caution: Note: Each mat includes an attached grounding snap. Mats must be grounded with a ground cord and workers must wear a heel grounder or shoes with conductive soles. Ground cords and heel grounders are available seperately from Wearwell.
Frequently Bought Together