EnzyChrom™ Glycerol Assay Kit, BioAssay Systems
Catalog # 75878-132
Supplier: BIOASSAY SYSTEMS
CAS Number:
Specifications
- Description:Glycerol Assay Kit
- Size:200 assays
- Method:OD570 nm, or FL530/585 nm
- Sample:Biological, food, beverage etc.
- Processing time:20 minutes
- Cat. no.:75878-132
- Supplier No.:EGLY-200
Specifications
About this item
For quantitative determination of glycerol and evaluation of drug effects on glycerol metabolism.
- Linear detection range 10 to 1000 μM (92 μg/dL to 9.2 mg/dL) glycerol for colorimetric assays and 2 to 50 μM for fluorimetric assays
- Homogeneous high-throughput assay
Glycerol [Glycerin or Glycerine, C3H5 (OH) 3] is widely used in foods, beverages and pharmaceutical formulations
BioAssay Systems’ glycerol assay uses a single Working Reagent that combines glycerol kinase, glycerol phosphate oxidase and color reactions in one step. The color intensity of the reaction product at 570nm or fluorescence intensity at λex/em = 530/585nm is directly proportional to glycerol concentration in the sample.
It is also a main by-product of biodiesel production. Simple, direct and automation-ready. Procedures for measuring glycerol concentrations find wide applications.