ContactUsLinkComponent
Safety-Solve Complete Counting Cocktail, RPI
  61000-504
 :  RPI
Small Business Enterprise
 :  
undefined
Safety-Solve Complete Counting Cocktail, RPI
  61000-504
 :  RPI
 :  111177
Small Business Enterprise
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Safety-Solve Complete Counting Cocktail
  • Size:
    4 L
  • Cat. no.:
    61000-504
  • Supplier No.:
    111177
Safety-Solve is a ready-to-use high flash point, cocktail for liquid scintillation counting.
This low-odor, high flash point, low viscosity formula does not require dilution prior to use. Contains a blend of high quantum efficiency quench resistant fluors in a non-ionic emulsifier system which will accommodate a wide range of samples.
Safety-solve complete counting cocktail is a bluish tint, liquid with slight odor.