ContactUsLinkComponent
QuantiChrom™ Bilirubin Assay Kit, BioAssay Systems
QuantiChrom™ Bilirubin Assay Kit, BioAssay Systems
Catalog # 75877-984
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
QuantiChrom™ Bilirubin Assay Kit, BioAssay Systems
Catalog # 75877-984
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  DIBR-180
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Bilirubin Assay Kit
  • Size:
    180 assays
  • Method:
    OD530 nm
  • Sample:
    Serum
  • Processing time:
    10 minutes
  • Cat. no.:
    75877-984
  • Supplier No.:
    DIBR-180
Specifications
About this item
For quantitative determination of bilirubin and evaluation of drug effects on bilirubin metabolism.
  • Detection limit of 0.16 mg/dL bilirubin in 96-well plate assay
  • Can be readily automated to process thousands of samples per day
Bilirubin is one of the degradation products of hemoglobin formed when red blood cells die
BioAssay Systems’ bilirubin assay kit is designed to measure bilirubin in blood specimen in 96-well or cuvette formats. The improved Jendrassik-Grof method utilizes the reaction of bilirubin with diazotized sulfanilic acid, in which a red colored product is formed. The intensity of the color, measured at 510-550nm, is an accurate measure of the bilirubin level in the sample. Total bilirubin is assessed using caffeine benzoate to split bilirubin from the unconjugated bilirubin protein complex.
Bilirubin exists in the insoluble unconjugated form (also indirect bilirubin), or soluble glucuronide conjugated form bilirubin (also direct bilirubin). Conjugated bilirubin moves into the bile canaliculi of the liver and then to the gall bladder. When stimulated by eating, bile (including the conjugated bilirubin) is excreted into the small intestine, where bilirubin is converted into urobilinogen. Bilirubin is a key diagnostic indicator. High levels of bilirubin result when too much hemoglobin is broken down or the removal of bilirubin does not function properly. The accumulation of bilirubin in the body causes jaundice. Simple and automation-ready. Procedures for quantitative determination of bilirubin find wide applications in research and drug discovery.