Vacuum Manifolds and Accessories for SPE, Honeywell Burdick
Catalogue # BJ9300
Fournisseur: Burdick & Jackson
Spécifications
- Description:16-Port Vacuum Manifold
- Cat. no.:BJ9300
- Supplier No.:9300
Spécifications
À propos de cet article
The exit port of the SPE columns is a Luer tip that allows the columns to be affixed securely to the vacuum manifold.
Delivery: Vacuum manifold includes cover, gasket, vacuum gauge, valve, glass block, 12 or 24 needles and stopcocks and collection racks (1 each size).