The Yankauer Pediatric Suction Tube is used during procedures on pediatric patients to clean away excess fluids, smoke, or debris from the surgical site.
- All Sklar products are industry standard
Both the tip size and overall length of the pediatric tube is smaller than the standard size Yankauer tube.
This removable tip is 8mm in diameter and 8.5" long overall. The tube is long and has two slight angles for ease of accessing the surgical site without obstructing the surgeon's view. The Yankauer pediatric suction tube is sometimes referred to as the Andrews-Pynchon suction tube.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 76192-894
- 10195-216
- Latex/Latex-free:
- Latex-free