À propos de cet article
These practical standard ovens have a maximum temperature of 300 °C.
- Digital PID Controller for Easy Constant Operation
- K Thermocouple Sensor
- Measured Temp., Temp Sensor., and Temp. Input Circuit Error Features
- Natural Convection Circulation Method
- Two Stainless Steel Plates with Four Brackets Provided
Natural convection ovens are economical and easy to use
The ovens feature a multitude of functions, stainless steel interiors, and an electro-galvanized steel sheet with melamine resin baking finish exteriors. Operation functions include a fixed-temperature operation, quick auto stop, and auto stop and start. A calibration offset, a power-failure compensation, and a key lock function are also included. Safety features include self-diagnostic functions, an hydraulic independent overheat prevention device, and electric leakage breaker with over-current protection.
The temperature setting can be digitally adjusted. Green LED lights display measurement temperatures, and red LED lights display setting temperatures. A SSR control adjusts the heater circuit. The exhaust port is made of two 33 mm pieces on the top of the ovens. The iron-chrome wire heater is 0.9 or 1.36 kW, depending on size.
Spécifications
- Catalog No:
- 75846-326
- 75846-330
- 75846-322
- Insulation:
- Glass Wool
- Glass Wool
- Glass Wool
- Max. load per shelf:
- 15 kg
- 15 kg
- 15 kg
- Timer:
- 1 Minute to 99 Hours, 59 Minutes and 100 Hours to 999 Hours, 50 Minutes
- 1 Minute to 99 Hours, 59 Minutes and 100 Hours to 999 Hours, 50 Minutes
- 1 Minute to 99 Hours, 59 Minutes and 100 Hours to 999 Hours, 50 Minutes