Corning® BioCoat™ Pre-Coated PAMPA Plate System, Corning
Catalog # 10018-852
Supplier: Corning
Specifications
- Description:96-Well pre-coated filter plate
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO 14001 - Bedford (reg. no. 14.4942)
- Cat. no.:10018-852
- Supplier No.:353015
Specifications
About this item
A 96-well, pre-coated filter plate, individually packaged with a 96-well microassay receiver plate.
- 96-well insert system with a filter plate pre-coated with structured layers of phospholipids
- Configuration complies with ANSI standards
- Polystyrene plate with 0.4 µm Polyvinylidene fluoride membrane