VeriSpec® Silicate - SiO₃²⁻ Standard for Ion Chromatography 1.000 +/- 0.002 g/L in NaOH 1%, RICCA Chemical Company
Supplier: Ricca Chemical
Certificates
About this item
Certified reference material (CRM) for ion chromatography (IC)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Frequently Bought Together