VeriSpec® Tungsten Standard for ICP/MS 100 ppm NH₄OH, RICCA Chemical Company
RCRV010565100N
:
- Concentration:100 µg/ml
- Matrix:H₂O/tr. NH₄OH
- Pack type:Plastic Bottle
- Size:100 ml
- Cat. no.:RCRV010565100N
- Element:Tungsten
- Supplier No.:RV010565-100N
Certified reference material (CRM) for ICP-MS
: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility