VeriSpec® Sulfur Standard for ICP/MS 100 ppm in H₂O, RICCA Chemical Company
Catalogue # RCRV010549100N
Fournisseur: Ricca Chemical
Numéro CAS:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Concentration:100 µg/ml
- Matrix:H₂O
- Pack type:Plastic Bottle
- Size:100 ml
- Cat. no.:RCRV010549100N
- Element:Sulfur
- Supplier No.:RV010549-100N
Spécifications
À propos de cet article
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility