VeriSpec® Selenium (Se) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Fournisseur: Ricca Chemical
Certificats
À propos de cet article
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Frequently Bought Together