StoreBox™ Storage Boxes, Simport
StoreBox™ Storage Boxes, Simport
StoreBox™ Storage Boxes, Simport
StoreBox™ Storage Boxes, Simport
These economical storage boxes are designed to be used at temperatures between –90 and 80 °C. Different models are available to accommodate either 25, 81, or 100 sample tubes.
  • Cover has numbered squares for easy sample identification
  • Two corners of cover and base are cut to prevent misalignment
  • Writing surface provided to identify base and/or cover
  • Vials readily visible through transparent cover
  • Air vents minimize condensation
  • Four pastel colors available for better color coding
A transparent cover allows the user to see the contents of the box. The cover is keyed to the base to prevent misalignment. Printed with a series of squares (numbered), the surface accepts writing with markers, facilitating inventory control.
A unique color coding system uses colored plastic grids to separate the cover from the base on the 25- and 81- place boxes. Those made to accept 100 tubes have a colored base instead of a grid. Vial removal is facilitated by an innovative vial picker supplied with each storage box. Not autoclavable.
Product Family Options
25-Place Storage Boxes
  • Désignation
    Color
    Matière
    Dimensions
  • StoreBox™ for 1 - 2 ml Tubes
    Blue
    PS
    3×3×21/6"
    Pack of 8
    Pack of 8 - $78.09
    $78.09
    Case of 48
    Case of 48 - $468.54
    $468.54
    Pack of 8
       $78.09
    Case of 48
       $468.54
     : T514-225B
  • StoreBox™ for 1 - 2 ml Tubes
    Blue
    PS
    3×3×21/6"
     : T514-225B
  • StoreBox™ for 1 - 2 ml Tubes
    Green
    PS
    3×3×21/6"
    Pack of 8
    Pack of 8 - $78.09
    $78.09
    Case of 48
    Case of 48 - $468.54
    $468.54
    Pack of 8
       $78.09
    Case of 48
       $468.54
     : T514-225G
  • StoreBox™ for 1 - 2 ml Tubes
    Green
    PS
    3×3×21/6"
     : T514-225G
  • StoreBox™ for 1 - 2 ml Tubes
    Pink
    PS
    3×3×21/6"
    Pack of 8
    Pack of 8 - $78.09
    $78.09
    Case of 48
    Case of 48 - $468.54
    $468.54
    Pack of 8
       $78.09
    Case of 48
       $468.54
     : T514-225P
  • StoreBox™ for 1 - 2 ml Tubes
    Pink
    PS
    3×3×21/6"
     : T514-225P
  • StoreBox™ for 1 - 2 ml Tubes
    Yellow
    PS
    3×3×21/6"
    Pack of 8
    Pack of 8 - $78.09
    $78.09
    Case of 48
    Case of 48 - $468.54
    $468.54
    Pack of 8
       $78.09
    Case of 48
       $468.54
     : T514-225Y
  • StoreBox™ for 1 - 2 ml Tubes
    Yellow
    PS
    3×3×21/6"
     : T514-225Y
81-Place Storage Boxes
  • Désignation
    Color
    Matière
    Dimensions
  • StoreBox™ for 1 - 2 ml Tubes
    Blue
    PS
    51/4×51/4×21/16"
    Pack of 4
    Pack of 4 - $57.28
    $57.28
    Case of 24
    Case of 24 - $343.68
    $343.68
    Pack of 4
       $57.28
    Case of 24
       $343.68
     : T514-281B
  • StoreBox™ for 1 - 2 ml Tubes
    Blue
    PS
    51/4×51/4×21/16"
     : T514-281B
  • StoreBox™ for 1 - 2 ml Tubes
    Green
    PS
    51/4×51/4×21/16"
    Pack of 4
    Pack of 4 - $57.54
    $57.54
    Case of 24
    Case of 24 - $345.24
    $345.24
    Pack of 4
       $57.54
    Case of 24
       $345.24
     : T514-281G
  • StoreBox™ for 1 - 2 ml Tubes
    Green
    PS
    51/4×51/4×21/16"
     : T514-281G
  • StoreBox™ for 1 - 2 ml Tubes
    Pink
    PS
    51/4×51/4×21/16"
    Pack of 4
    Pack of 4 - $57.76
    $57.76
    Case of 24
    Case of 24 - $346.56
    $346.56
    Pack of 4
       $57.76
    Case of 24
       $346.56
     : T514-281P
  • StoreBox™ for 1 - 2 ml Tubes
    Pink
    PS
    51/4×51/4×21/16"
     : T514-281P
  • StoreBox™ for 1 - 2 ml Tubes
    Yellow
    PS
    51/4×51/4×21/16"
     : -
  • StoreBox™ for 1 - 2 ml Tubes
    Yellow
    PS
    51/4×51/4×21/16"
     : T514-281Y
  • StoreBox™ for 3 - 5 ml Tubes
    Blue
    PS
    51/4×51/4×33/4"
    Pack of 5
    Pack of 5 - $164.40
    $164.40
    Case of 10
    Case of 10 - $328.80
    $328.80
    Pack of 5
       $164.40
    Case of 10
       $328.80
     : T514-581B
  • StoreBox™ for 3 - 5 ml Tubes
    Blue
    PS
    51/4×51/4×33/4"
     : T514-581B
  • StoreBox™ for 3 - 5 ml Tubes
    Green
    PS
    51/4×51/4×33/4"
    Pack of 5
    Pack of 5 - $166.61
    $166.61
    Case of 10
    Case of 10 - $333.22
    $333.22
    Pack of 5
       $166.61
    Case of 10
       $333.22
     : T514-581G
  • StoreBox™ for 3 - 5 ml Tubes
    Green
    PS
    51/4×51/4×33/4"
     : T514-581G
  • StoreBox™ for 3 - 5 ml Tubes
    Pink
    PS
    51/4×51/4×33/4"
    Pack of 5
    Pack of 5 - $166.60
    $166.60
    Case of 10
    Case of 10 - $333.20
    $333.20
    Pack of 5
       $166.60
    Case of 10
       $333.20
     : T514-581P
  • StoreBox™ for 3 - 5 ml Tubes
    Pink
    PS
    51/4×51/4×33/4"
     : T514-581P
  • StoreBox™ for 3 - 5 ml Tubes
    Yellow
    PS
    51/4×51/4×33/4"
    Pack of 5
    Pack of 5 - $166.61
    $166.61
    Case of 10
    Case of 10 - $333.22
    $333.22
    Pack of 5
       $166.61
    Case of 10
       $333.22
     : T514-581Y
  • StoreBox™ for 3 - 5 ml Tubes
    Yellow
    PS
    51/4×51/4×33/4"
     : T514-581Y
100-Place Storage Boxes
  • Désignation
    Color
    Matière
    Dimensions
  • StoreBox™ for 1 - 2 ml Tubes
    Blue
    PS
    51/4×51/4×21/16"
    Pack of 4
    Pack of 4 - $54.86
    $54.86
    Case of 24
    Case of 24 - $329.16
    $329.16
    Pack of 4
       $54.86
    Case of 24
       $329.16
     : T514-2100B
  • StoreBox™ for 1 - 2 ml Tubes
    Blue
    PS
    51/4×51/4×21/16"
     : T514-2100B
  • StoreBox™ for 1 - 2 ml Tubes
    Green
    PS
    51/4×51/4×21/16"
    Pack of 4
    Pack of 4 - $54.61
    $54.61
    Case of 24
    Case of 24 - $327.66
    $327.66
    Pack of 4
       $54.61
    Case of 24
       $327.66
     : T514-2100G
  • StoreBox™ for 1 - 2 ml Tubes
    Green
    PS
    51/4×51/4×21/16"
     : T514-2100G
  • StoreBox™ for 1 - 2 ml Tubes
    Pink
    PS
    51/4×51/4×21/16"
    Pack of 4
    Pack of 4 - $54.61
    $54.61
    Case of 24
    Case of 24 - $327.66
    $327.66
    Pack of 4
       $54.61
    Case of 24
       $327.66
     : T514-2100P
  • StoreBox™ for 1 - 2 ml Tubes
    Pink
    PS
    51/4×51/4×21/16"
     : T514-2100P
  • StoreBox™ for 1 - 2 ml Tubes
    Yellow
    PS
    51/4×51/4×21/16"
     : -