Prosolv® Recycling System and Components, Justrite® | Avantor
Prosolv® Recycling System and Components, Justrite®
Supplier:   Justrite
Prosolv® Recycling System and Components, Justrite®
Supplier:  Justrite
Certificates
Prosolv® Recycling System and Components, Justrite®
2 Options Available Below
Product Details & Documents
Safely and economically convert gas cylinders into recyclable steel containers.
  • Prosolv® System removes the sealing sleeve and internal valve stem from propane, MAPP, and 0.75" male threaded calibration gas cylinders
  • Empties and filters propellants from cylinders, converting them into non-hazardous recyclable material
  • Not for use with oxygen bottles
Spent propane canisters are considered hazardous waste while still under pressure, but once they are safely vented and relieved of pressure they can be recycled as scrap steel. The Prosolv® System safely recycles depressurized propane cylinders, eliminating an entire category of hazardous waste from the waste stream.
Certifications: EPA Compliant.
Informations sur les accessoires: System includes vent device, carbon filter, antistatic wire, safety goggles, sleeve removal tool, and 40 certification tags.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Prosolv® Gas Cylinder Recycling System
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,435.40
    $2,435.40
    Prix promotionnel pour les nouveaux clients
    Each
       $2,435.40
    La quantité minimale de commande est de 1
    Supplier Number: 28190
  • Prosolv® Replacement Filter with 40 Tags
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $557.05
    $557.05
    Prix promotionnel pour les nouveaux clients
    Each
       $557.05
    La quantité minimale de commande est de 1
    Supplier Number: 28191