Rugged Elmasonic xtra ST high capacity ultrasonic cleaners are designed for cleaning a wide range of parts in a production environment.
- Powerful heavy-duty ultrasonic cleaner
- Portable and easy to use
- Dual frequency for coarse and fine cleaning
- Sweep mode evenly distributes cleaning
- Pulse mode adds bursts of ultrasonic power for intensive cleaning and degassing
- Dynamic mode combines Sweep and Pulse for optimum cleaning results
Stubborn contaminants like polishing pastes are easily removed at 25 kHz
The stainless steel tanks are wheel mounted for portability. The timer can be set from 1 to 30 minutes or for continuous operation. Temperature is adjustable from 30˚C to 80˚C with automatic start up when the set temperature is reached. For safety purposes the unit automatically shuts off at 90˚C or after 12 hours of continuous operation. All models have drains with stainless steel ball valves.
The xtra ST sonic cleaner is designed to be easily serviced with minimum down time. The control panel/generator assembly can be quickly removed and replaced in house. Intelligent generators adjust themselves automatically to the transducers.
Elmasonic xtra ST devices are rated for continuous operation under the highest operational demands. The robust tanks made of special stainless steel have a long service life. Dependable state-of-the-art Elma ultrasonic cleaners are made in Germany.
- Catalog No:
- 75848-816
- 75848-818
- 75848-820
- 75848-810
- 75848-764
- 75848-812
- 75848-814
- Environmentally Preferable:
- Sustainable Packaging, Safer Human and Environmental Health
- Sustainable Packaging, Safer Human and Environmental Health
- Sustainable Packaging, Safer Human and Environmental Health
- Sustainable Packaging, Safer Human and Environmental Health
- Sustainable Packaging, Safer Human and Environmental Health
- Sustainable Packaging, Safer Human and Environmental Health
- Sustainable Packaging, Safer Human and Environmental Health
- Frequency:
- 25 and 45 kHz
- 25 and 45 kHz
- 25 and 45 kHz
- 25 and 45 kHz
- 25 and 45 kHz
- 25 and 45 kHz
- 25 and 45 kHz
- Temperature range:
- 30°C to 80°C
- 30°C to 80°C
- 30°C to 80°C
- 30°C to 80°C
- 30°C to 80°C
- 30°C to 80°C
- 30°C to 80°C
- Timer:
- 1-30 minutes or continuous
- 1-30 minutes or continuous
- 1-30 minutes or continuous
- 1-30 minutes or continuous
- 1-30 minutes or continuous
- 1-30 minutes or continuous
- 1-30 minutes or continuous
- Safety:
- Automatic shut down after 12 hours of continuous operation or when temperature reaches 90°C
- Automatic shut down after 12 hours of continuous operation or when temperature reaches 90°C
- Automatic shut down after 12 hours of continuous operation or when temperature reaches 90°C
- Automatic shut down after 12 hours of continuous operation or when temperature reaches 90°C
- Automatic shut down after 12 hours of continuous operation or when temperature reaches 90°C
- Automatic shut down after 12 hours of continuous operation or when temperature reaches 90°C
- Automatic shut down after 12 hours of continuous operation or when temperature reaches 90°C