7000 Series Reusable Half Mask Respirators, Moldex®
Supplier: Moldex/Metric
Certificates
About this item
These 7000 series half mask respirators are designed to comfortably conform to facial contours and with a modern look that increases worker acceptance
- 100% PVC-Free®
- Suited to be worn under welding helmets, wide field of vision
- Higher resistance to heat and compression
- Enhanced comfort and durability of silicone
- Adjustable head cradle and curved neck buckles for extra comfort
Sleek design for modern, high-tech look and increased worker acceptance. Light weight design plus extra-wide sealing area provides all-day comfort. Low profile, compact design fits well under welding helmets and with safety glasses. Mask drops down for convenient storage around the neck or locks down for custom fit. Available pre-assembled and also as individual components
The curved neck buckle and adjustable head cradle ensure a comfortable, custom fit for most users and without pressure points.
Specifications
- Catalog No:
- 10065-314
- 10065-316
- 10065-318
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health