À propos de cet article
Round metal specimen chuck with O-ring made of low temperature resistant rubble; with stem length ~37.5 mm.
- Different o-ring colors eliminates sample errors and the use of expensive colored freezing medium
- Orientation mark to assure proper sample positioning
Proper dimensional tolerance and soft aluminum, to avoid slippage slip in chuck that results in thin/thick cutting.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.