TempDex 710, Nitrile Gloves, Mapa Professional
 :   MAPA
TempDex 710, Nitrile Gloves, Mapa Professional
 :  MAPA
TempDex 710, Nitrile Gloves, Mapa Professional
TempDex 710, Nitrile Gloves, Mapa Professional
These high dexterity and thermal protection gloves are suitable for handling of composite materials after heat processing in ceramics, plastics and composite industries.
  • Nitrile coated palm and dotted embossing for better insulation and grip
  • Thin, anatomically-shaped glove ensures dexterity
  • Long lasting and abrasion resistant
Ideal for use in ceramics, plastics, composite and construction industries.
Product Family Options
  • Taille du gant
    Glove Color
    Longueur
  • 7
    Yellow
    26 cm (10³⁄₁₆")
    Each
    Each - $19.19
    $19.19
    Case of 50
    Case of 50 - $959.50
    $959.50
    Each
       $19.19
    Case of 50
       $959.50
     : 34710037
  • 9
    Yellow
    26 cm (10³⁄₁₆")
    Each
    Each - $13.82
    $13.82
    Case of 50
    Case of 50 - $691.23
    $691.23
    Each
       $13.82
    Case of 50
       $691.23
     : 34710039
  • 11
    Yellow
    26 cm (10³⁄₁₆")
     : -