VWR® Balance Series Sample Holders
À propos de cet article
VWR® Sample holders are an ideal solution to aid in faster and more convenient weighing of laboratory products.
Sample holders are a great tool for facilitating dosing directly into typical laboratory vessels.
Sample holders are compatible with the VWR® A-Series balances (Analytical balances with draft shield: 10205-026, 10204-960, 10204-962, and 10204-964).
4 Options Available Below
Product Details & Documents
VWR® Sample holders are an ideal solution to aid in faster and more convenient weighing of laboratory products.
Sample holders are a great tool for facilitating dosing directly into typical laboratory vessels.
Sample holders are compatible with the VWR® A-Series balances (Analytical balances with draft shield: 10205-026, 10204-960, 10204-962, and 10204-964).