Temperature Controller with Process and Limit Control, Ace Glass
À propos de cet article
Temperature controller combines a Process control and a Limit control in one box.
- LED is user-selectable for Fahrenheit or Centigrade
Controller has two 900 watt outlets, independent Process and Limit sensors, and audible alarms. The Limit control will turn off the power when a user-selectable value is reached. Controller requires a manual reset if tripped.
Certifications: cULus rated.
Ordering information: Includes integral rack mounting bracket. One-year limited manufacturer’s warranty.
6 Options Available Below
Product Details & Documents
Temperature controller combines a Process control and a Limit control in one box.
- LED is user-selectable for Fahrenheit or Centigrade
Controller has two 900 watt outlets, independent Process and Limit sensors, and audible alarms. The Limit control will turn off the power when a user-selectable value is reached. Controller requires a manual reset if tripped.
Certifications: cULus rated.
Ordering information: Includes integral rack mounting bracket. One-year limited manufacturer’s warranty.
Specifications for Product Family
Spécifications
- Catalog No:
- 75846-476
- 75846-478
- 75846-488
- 75846-490
- 75846-492
- 75846-494
- Accuracy:
- 0.1% of span, ±1.0 °C, with setpoint ramping feature
- 0.1% of span, ±1.0 °C, with setpoint ramping feature
- 0.1% of span, ±1.0 °C, with setpoint ramping feature
- 0.1% of span, ±1.0 °C, with setpoint ramping feature
- 0.1% of span, ±1.0 °C, with setpoint ramping feature
- 0.1% of span, ±1.0 °C, with setpoint ramping feature
- Electrical:
- 120VAC, 15A, 60Hz input
- 120VAC, 15A, 60Hz input
- 120VAC, 15A, 60Hz input
- 120VAC, 15A, 60Hz input
- 120VAC, 15A, 60Hz input
- 120VAC, 15A, 60Hz input
Spécifications
Product Family Options
Informations sur le produit
- DésignationIncludesLead TypeProbe TypeAvailabilityPrice
- Dual 900 Watt Outlet Temperature Control with Process and Limit Control and Audible Alarms——JLog in pour la disponibilitéEach$5,871.53Supplier Number: 12336-10Spécifications:
More
Specifications
Cat. No.75846-476Accuracy0.1% of span, ±1.0 °C, with setpoint ramping featureElectrical120VAC, 15A, 60Hz input - Dual 900 Watt Outlet Temperature Control with Process and Limit Control and Audible Alarms1/4 x 12" Inconel Sheathed ProbeDual 72" SMP leadsJLog in pour la disponibilitéEach$6,273.79Supplier Number: 12336-15Spécifications:
More
Specifications
Cat. No.75846-478Accuracy0.1% of span, ±1.0 °C, with setpoint ramping featureElectrical120VAC, 15A, 60Hz input - Dual 900 Watt Outlet Temperature Control with Process and Limit Control and Audible Alarms——KLog in pour la disponibilitéEach$5,695.83Supplier Number: 12336-20Spécifications:
More
Specifications
Cat. No.75846-488Accuracy0.1% of span, ±1.0 °C, with setpoint ramping featureElectrical120VAC, 15A, 60Hz input - Dual 900 Watt Outlet Temperature Control with Process and Limit Control and Audible Alarms1/4 x 12" Inconel Sheathed ProbeDual 72" SMP leadsKLog in pour la disponibilitéEach$6,117.10Supplier Number: 12336-25Spécifications:
More
Specifications
Cat. No.75846-490Accuracy0.1% of span, ±1.0 °C, with setpoint ramping featureElectrical120VAC, 15A, 60Hz input - Dual 900 Watt Outlet Temperature Control with Process and Limit Control and Audible Alarms——TLog in pour la disponibilitéEach$5,871.53Supplier Number: 12336-30Spécifications:
More
Specifications
Cat. No.75846-492Accuracy0.1% of span, ±1.0 °C, with setpoint ramping featureElectrical120VAC, 15A, 60Hz input - Dual 900 Watt Outlet Temperature Control with Process and Limit Control and Audible Alarms1/4 x 12" Inconel Sheathed ProbeDual 72" SMP leadsTLog in pour la disponibilitéEach$6,347.56Supplier Number: 12336-35Spécifications:
More
Specifications
Cat. No.75846-494Accuracy0.1% of span, ±1.0 °C, with setpoint ramping featureElectrical120VAC, 15A, 60Hz input