À propos de cet article
Histology Embedding Rings provide a quick and easy way to uniformly embed tissues for further processing.
- Uniform Block Size For Sectioning
- Work With Stainless Steel Or Plastic Base Molds
- Provide Convenient Storage And Identification
Simply place the embedding ring atop a base mold, fill with paraffin, and allow to cool
Top quality, made in the USA.
Available in Three colors for color coding references.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.