Difco™ Legionella Agar Enrichment, BD Diagnostics
CA90002-656
: BD
:
- Description:Legionella Agar Enrichment
- Size:5 mL
- Cat. no.:CA90002-656
- Supplier No.:233901
Legionella Agar Enrichment is intended for use in the the preparation of Legionella Agar.
- Based on the Charcoal Yeast Extract (CYE) Agar formula
- Recommended for use in the isolation and cultivation of Legionella from clinical and non-clinical materials
- Complete medium
Legionella Agar Enrichment is intended for use in the the preparation of Legionella Agar. The complete medium is based on the Charcoal Yeast Extract (CYE) Agar formula of Feeley et al., as modified by Edelstein, and is recommended for use in the isolation and cultivation of Legionella from clinical and non-clinical materials.