Support Extension, Glas-Col®, LLC
Fournisseur: Glas-Col
Certificats
À propos de cet article
Table top support for rounded bottom fabric mantles.
- Steel support ring
- Fabric strap basket cradle
- Durable construction
- For use with round bottom soft shell mantle
Our table top support holds spherical and hemispherical mantles up to 5 L. Attaches easily to ring stands or distillation racks.
Delivery: 7 to 10 days after order.
Frequently Bought Together