Support Tripod, Glas-Col®, LLC
Support Tripod, Glas-Col®, LLC
 :   Glas-Col
Support Tripod, Glas-Col®, LLC
 :  Glas-Col
Support Tripod, Glas-Col®, LLC
Support Tripod, Glas-Col®, LLC
Floor height support for rounded bottom aluminum shell mantles.
  • Stable three leg design
  • Floor or bench mount
  • Durable construction
Our tripod support sits on the floor or bench to hold hard shell rounded bottom mantles ranging in sizes from 3 L to 72 L. Mounting holes are pre drilled in each leg.
Product Family Options
  • Désignation
    Weight
  • Tripod, 3 L capacity
    4 lbs.
    Each
    Each - $435.60
    $435.60
    Each
       $435.60
     : 100EM505
  • Tripod, 5 L capacity
    4 lbs.
    Each
    Each - $449.17
    $449.17
    Each
       $449.17
     : 100EM510
  • Tripod, 12 L capacity
    8 lbs.
    Each
    Each - $558.11
    $558.11
    Each
       $558.11
     : 100EM515
  • Tripod, 22 L capacity
    8 lbs.
    Each
    Each - $756.77
    $756.77
    Each
       $756.77
     : 100EM520
  • Tripod, 50 L capacity
    11 lbs.
    Each
    Each - $696.20
    $696.20
    Each
       $696.20
     : 100EM525
  • Tripod, 72 L capacity
    12 lbs.
    Each
    Each - $814.22
    $814.22
    Each
       $814.22
     : 100EM530