Heating Mantles, High Temp, Glas-Col®
Fournisseur: Glas-Col
Certificats
À propos de cet article
Aluminum housed mantle for applications requiring more heat.
- Fabric interior
- Durable aluminum outer shell
- Adaptable to most vessels
The Series "STM" mantle is designed for applications requiring a higher temperature. The interior is resilient glass fabric which nesltles the glassware. The exterior is rigid aluminum providing strength and support. The mantle has an internal operating temp of up to 600 degrees C.
Delivery: 7-10 days after order.
Spécifications
- Catalog No:
- 89260-004
- 89260-002
- 33752-450
- Electrical:
- 115V
- 115V
- 115V