Lab Stand, Glas-Col
Lab Stand, Heavy Duty, Up to 16" dia. Containers
Catalog # 58954-025
Supplier: Glas-Col
Fournisseur #: 099DA130150
Jump to:
Product Details & Documents
Heavy duty lab stand made to last with stainless steel mast and cast iron base.
- Sturdy construction
- Accepts up to 16" diameter containers
- Heavy tip resistant base
This heavy duty lab stand will help to prevent tipping.The balancing weight of the cast iron base and the 3/4" diameter mast make it ideal for use with our stirrers, stir testers and homogenizers.
Delivery information: 7-10 days after order.
Spécifications
- Désignation:Lab Stand, Heavy Duty
- For Use With:Up to 16" dia. Containers