VWR® Repeating Dispenser and Syringe Tips
Fournisseur: VWR International
Certificats
À propos de cet article
Designed for quick, accurate, and easy repeat pipetting. The smooth-moving dosing button and lightweight design facilitates long dosing series without causing strain or discomfort to the user.
- Lightweight, ergonomic design
- Allows for 9 tip sizes, 10 volume adjustments, and 90 program steps
Nine different tip sizes are available, for dispensing volumes ranging from 1 μl to 50 ml. The device provides reliable and repeatable results for a wide range of liquids, including viscous, infectious, and volatile samples. Available in sterile and nonsterile options. All sterile tips are individually wrapped.
Volume selection, loading, and dispensing of liquids can be performed with ease using only one hand.
Certifications: All sterile tips are certified to be pyrogen-, ATP-, RNase-, and DNase-free. The VWR® Repeating Dispenser is manufactured under DIN EN ISO 8655 requirements using high-quality materials, ensuring a device that is built to last.
VWR® Classic Syringe Tips, Nonsterile