ContactUsLinkComponent
Brucella Pea And KVL OxyPRAS Plus Multi-Pack, Oxyrase, Inc.
Brucella Pea And KVL OxyPRAS Plus Multi-Pack, Oxyrase, Inc.
Catalog # 101975-897
Supplier:  Oxyrase Inc
Brucella Pea And KVL OxyPRAS Plus Multi-Pack, Oxyrase, Inc.
Catalog # 101975-897
Supplier:  Oxyrase Inc
Supplier Number:  M-BKP-3
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Size:
    Pack of 10
  • Cat. no.:
    101975-897
  • Storage Temperature:
    +2…+8 °C
  • Supplier No.:
    M-BKP-3
Specifications
About this item
Individually packaged BRU, PEA and KVL plates; banded together. OxyPRAS® Plates are specifically made to improve recovery of anaerobes.
  • Specifically made for anaerobes to improve recovery of injured cells
  • Arrive reduced and ready to use
  • Stays reduced in air for hours
  • Saves tech time and helps eliminate errors
  • Specially packaged in oxygen barrier bags
  • Store refrigerated for a extended shelf life or at room temperature
  • Meets Best Practice criteria for working with clinical anaerobes
Due to the Oxyrase® Enzyme System, OxyPRAS Plus® Plates (Pre-Reduced Anaerobically Sterilized) are technologically advanced compared to other plates.
Oxyrase® utilized during the manufacture of the plate prevents oxidation during its preparation. Oxyrase® is again added to the final plate. Oxyrase® keeps the plate reduced and confers unique properties to the OxyPRAS Plus® plate not found in standard or conventional PRAS plates. PRAS plates are considered the gold standard to isolate all anaerobes from clinical specimens.
OxyPRAS Plus® Multi-Packs are individually wrapped plates; banded together. OxyPRAS Plus® Multi-Packs save time, help eliminate errors, add value and flexibility by being banded together. Each plate has its own lot number. Six month shelf-life when refrigerated and three month expiration date at room temperature.