Drip Pads, Justrite®
 :   Justrite
Drip Pads, Justrite®
 :  Justrite
Drip Pads, Justrite®
Drip Pads, Justrite®
Capture small oil leaks and drips before they contaminate the ground or create unsafe work surfaces.
  • Lightweight, flexible, and portable - great for containing small oil drips from leaky vehicles, industrial machinery equipment, hydraulic lines, pipes, and drums.
  • Made from UV- and chemical-resistant modified PVC.
  • Patented design feature includes a coated mesh outer layer to trap coarse debris.
These drip pads feature replaceable, oil-only sorbent with a welded leakproof barrier. Rated for drive over applications, pads include brass grommets in opposing corners for anchoring. Anchoring stakes not included. Pads come in two sizes; extended model is weighted to keep it down in high winds. Outer shell is reusable with optional refill sorbent packs. Helps comply with EPA for spill prevention (SPCC).
Product Family Options
  • Désignation
    Exterior Dimensions
    Capacité d'absorption
    Weight
  • Pad Drip Standard
    21 W x 23 D"
    5 L
    2 lbs.
    Each
    Each - $256.36
    $256.36
    Each
       $256.36
     : 28458
  • Pad Drip Extended
    24 W x 36 D"
    7 L
    7 lbs.
    Each
    Each - $387.58
    $387.58
    Each
       $387.58
     : 28459
  • Pad Drip Extended Refills
    24 W x 36 D"
    7 L
    7 lbs.
    Each
    Each - $138.63
    $138.63
    Each
       $138.63
     : 28461
  • Pad Drip Refills
    21 W x 23 D"
    5 L
    2 lbs.
    Each
    Each - $49.66
    $49.66
    Each
       $49.66
     : 28460