Washdown Quickberm, Justrite®
Supplier:   Justrite
Washdown Quickberm, Justrite®
Supplier:  Justrite
Certificates
Washdown Quickberm, Justrite®
Washdown Quickberm, Justrite®
3 Options Available Below
Product Details & Documents
Washdowns use reinforced fabric material to withstand drive-thru traffic, including backhoes and skid steers.
  • Two 1" NPT fittings on each side of the berm for drainage
  • Simply remove foam inserts to fold material for quick and compact storage
Durable EnGuard™V, modified PVC coated fabric material reinforced with radio frequency-welded, high visibility yellow runway strips that withstand drive-thru traffic, including backhoes and skid steers.
Product Family Options
Informations sur le produit
  • Désignation
    Interior Dimensions
    Exterior Dimensions
    Availability
    Price
  • Quickberm Washdown
    120 W x 4 H x 132 D"
    128 W x 4 H x 168 L"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (250US Gal)
    Each (250US Gal) - $4,865.23
    $4,865.23
    Prix promotionnel pour les nouveaux clients
    Each (250US Gal)
       $4,865.23
    Supplier Number: 28576
  • Quickberm Washdown
    120 W x 4 H x 216 D"
    128 W x 4 H x 252 L"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (425US Gal)
    Each (425US Gal) - $5,976.13
    $5,976.13
    Each (425US Gal)
       $5,976.13
    Supplier Number: 28578
  • Quickberm Washdown
    168 W x 4 H x 648 L"
    176 W x 4 H x 680 L"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (1,800US Gal)
    Each (1,800US Gal) - $12,671.84
    $12,671.84
    Each (1,800US Gal)
       $12,671.84
    Supplier Number: 28581