QuickBerm Lite Containment Berm, Justrite®
À propos de cet article
Portable, versatile containment berm deploys quickly for emergency spill response.
- Folds down easily to a small size for convenient storage
- Complies with EPA for containment and spill prevention
Lightweight berm folds down easily to a small size for storage in trucks, emergency vehicles, and spill kits. Add optional grates to help reduce contact with contaminated liquids by personnel or equipment. Helps comply with EPA for containment and spill prevention (SPCC).
4 Options Available Below
Product Details & Documents
Portable, versatile containment berm deploys quickly for emergency spill response.
- Folds down easily to a small size for convenient storage
- Complies with EPA for containment and spill prevention
Lightweight berm folds down easily to a small size for storage in trucks, emergency vehicles, and spill kits. Add optional grates to help reduce contact with contaminated liquids by personnel or equipment. Helps comply with EPA for containment and spill prevention (SPCC).
Product Family Options
Informations sur le produit
- DésignationInterior DimensionsExterior DimensionsAvailabilityPrice
- Quickberm Lite48 W x 8 H x 48 D"48 W x 8 H x 48 L"Log in pour la disponibilitéLog in pour la disponibilitéEach (79US Gal)Each (79US Gal) - 681,88 $681,88 $Prix promotionnel pour les nouveaux clientsEach (79US Gal)681,88 $Supplier Number: 28556
- Quickberm Lite48 W x 8 H x 72 L"48 W x 8 H x 72 L"Log in pour la disponibilitéLog in pour la disponibilitéEach (119US Gal)Each (119US Gal) - 1 655,56 $1 655,56 $Prix promotionnel pour les nouveaux clientsEach (119US Gal)1 655,56 $Supplier Number: 28558
- Quickberm Lite96 W x 8 H x 96 D"96 W x 8 H x 96 L"Log in pour la disponibilitéLog in pour la disponibilitéEach (318US Gal)Each (318US Gal) - 1 418,15 $1 418,15 $Prix promotionnel pour les nouveaux clientsEach (318US Gal)1 418,15 $Supplier Number: 28566
- Quickberm Lite96 W x 8 H x 120 D"96 W x 8 H x 120 L"Log in pour la disponibilitéLog in pour la disponibilitéEach (398US Gal)Each (398US Gal) - 1 534,59 $1 534,59 $Prix promotionnel pour les nouveaux clientsEach (398US Gal)1 534,59 $Supplier Number: 28568