pH Buffer Solutions, Color-Coded, LaMotte
pH Buffer Solutions, Color-Coded, LaMotte
Supplier:   LaMotte
pH Buffer Solutions, Color-Coded, LaMotte
Supplier:  LaMotte
Certificates
pH Buffer Solutions, Color-Coded, LaMotte
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Minute amount of color permits immediate visual distinction of different buffer values.
Product Family Options
Product Information
  • Taille
    pH
    Color
    Availability
    Price
  • 500 mL
    4.01
    Red
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $52.58
    $52.58
    Each (500ml)
       $52.58
    Supplier Number: 3771-L
  • 500 mL
    7.00
    Yellow
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $49.99
    $49.99
    Each (500ml)
       $49.99
    Supplier Number: 3772-L
  • 500 mL
    10.00
    Blue
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $49.99
    $49.99
    Each (500ml)
       $49.99
    Supplier Number: 3773-L