Cloud Detective Device
Cloud Detective Device, Pack of 5
  470221-234
 :  14360
Cloud Detective Device
  470221-234
Cloud Detective Device
Cloud Detective Device
Cloud Detective Device, Pack of 5
$33.20
Each Pack of 5
Observe the sky and identify various cloud types with this simple device.
  • Promote outdoor learning
  • Investigate the natural world
  • Additional activities available to download
Students can easily follow the provided instructions to identify the cloud types. Color, shape and weather conditions are printed in the back of the dial to help students identify the appropriate cloud type. Extension activities can be accessed by scanning the QR code or provided download link.
  • Désignation:
    Cloud Detective Device, Pack of 5