Acid Neutralization Trap, Welch® | Avantor
Acid Neutralization Trap, Welch®
Acid Neutralization Trap, 8×13"
Catalog # 54994-092
Supplier:   Welch Vacuum
Fournisseur #:  1420H-20
Acid Neutralization Trap, Welch®
Catalog # 54994-092
Supplier:  Welch Vacuum
Acid Neutralization Trap, Welch®
Acid Neutralization Trap, Welch®
Acid Neutralization Trap, 8×13"
Price
$1,091.60
Log in pour la disponibilité
Each
Product Details & Documents
The Acid Neutralization Trap contains a large alkaline element cartridge to neutralize hydrous acids before they enter the vacuum pump
  • ISO NW flange connection
  • Calcium Hydroxide element
  • Configure with prior cold trap to reduce loading
The element changes from white to bluish-transparent when spent and can be observed through the transparent trap body.
Spécifications
  • Désignation:
    Acid Neutralization Trap, 8×13"