ZWITTERGENT® 3-12 Detergent, Calbiochem®, MilliporeSigma
Supplier: MilliporeSigma
Certificates
About this item
This synthetic zwitterionic detergent retains its zwitterionic character over a broad pH range.
- Synonyms: n-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
- Form: White solid
- Storage: +15°C to +30°C
- Toxicity: Corrosive
- CMC: 2 - 4 mM
- Aggregation Number: 55
The zwitterionic character is attributed to the presence of both a strongly basic quaternary ammonium ion and an acidic sulfonate ion of equal strength. This ionic balance presumably prevents irreversible binding to either anionic or cationic compounds.
Caution: Following reconstitution, refrigerate (4 °C). Stock solutions are stable for up to 1 month at 4 °C.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together