ContactUsLinkComponent
Staticmaster® 1U400 Ionizing Cartridge, 1", NRD
Catalog # 75794-964
Supplier:  NRD Staticmaster
undefined
Staticmaster® 1U400 Ionizing Cartridge, 1", NRD
Catalog # 75794-964
Supplier:  NRD Staticmaster
Supplier Number:  1U400
Specifications
  • Description:
    Staticmaster® 1U400 ionizing cartridge, range 0.5 to 1.5" (optimum)
  • Emission:
    Alpha (Polonium-210 Isotope, Activity 500 µCi)
  • Cat. no.:
    75794-964
  • W×D×H:
    33x4.6x23.6 mm 1.3x0.18x0.93"
  • Weight:
    5.7 g (0.2 oz.)
  • Supplier No.:
    1U400
Specifications
About this item
The Staticmaster® 1U400 Ionizing Cartridge is a self-powered static eliminator with a small profile, making it an ideal solution for in-balance elimination
  • Self-powered
  • Small profile
  • Easy mounting
  • Ensures accurate weighing
  • 302 stainless steel housing and aluminum grid
This small profile ionizer offers "zero-volts balance" technology, which means that it can effectively remove all of the static charge present in your media and samples. This ensures accurate weighing every time.