À propos de cet article
CELLvo™ Matrix is a ready-to-use cell culture substrate which provides a native microenvironment that replicates the 3-dimensional "home" in which stem cells naturally reside and proliferate.
- CELLvo™ Matrix is a natural extracellular matrix synthesized in vitro by bone marrow stromal cells and includes the biochemistry and architecture normally found in the bone marrow
- CELLvo™ Matrix is an advanced stem cell culture system that replicates the 3-dimensional "home" in which stem cells naturally reside and proliferate
- Mesenchymal stem cells grown on CELLvo™ Matrix proliferate more rapidly
- Mesenchymal stem cells grown on CELLvo™ Matrix maintain a smaller cell size
- Mesenchymal stem cells grown on CELLvo™ Matrix exhibit a higher percentage of cells expressing SSEA-4
CELLvo™ Matrix is a ready-to-use cell culture substrate which provides a native microenvironment that replicates the 3-dimensional "home" in which stem cells naturally reside and proliferate.
CELLvo™ Matrix is a protein extracellular matrix that mimics the normal microenvironment of stem cells. This 3-D matrix is produced by young adult human bone marrow cells. The final product is a thin, off-white film on cell culture flasks, dishes, and substrates.
CELLvo™ Matrix is not for human use. This product should not be used for diagnostic or therapeutic purposes.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.