VWR® Basic Melting Point Apparatus
VWR® Basic Melting Point Apparatus
   Sustainable Options Available
Supplier:   VWR International
VWR® Basic Melting Point Apparatus
Supplier:  VWR International
   Sustainable Options Available
Certificates
VWR® Basic Melting Point Apparatus
2 Options Available Below
Product Details & Documents
The VWR® Basic Melting Point Apparatus is primarily used in determining the purity of a sample and is optimized to quickly and efficiently heat up to 3 samples at one time.
  • LCD screen displays temperature, date, and time
  • Audible alarm when set temperature is reached
  • Large, clear viewing window with 8x magnification and white LED to illuminate samples
  • Five-Year Warranty
  • Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
Microprocessor controlled units maintain precise temperature control from ambient +5 °C to 400 °C
Audible alarm will sound when the set temperature is reached. Safety features include an indefinite heating time out. If ramping phase is not initiated within 30 minutes of reaching the set temperature, the unit returns to standby mode and the heater turns off to prevent indefinite heating. To prevent overheating, there is a maximum temperature timeout. After 30 minutes at the maximum temperature, the unit reverts back to the set temperature to prevent overheating.
Unit includes integral holder for capillary tubes.
Easy-to-use controls allow temperature adjustments in 1 °C increments. Touch pad controls with LCD display for temperature, date, and time allow operator to view settings efficiently while monitoring samples through the viewing window. Fixed ramp rate from set temperature of 2 °C per minute. Provides repeatable and accurate results every time and is easily visible across lab benches. LED lamp illuminates the oven while the oven window provides 8x magnification of samples at an ergonomic viewing angle.
Certifications: TUV Listed RoHS Compliant.
Ordering information: All units include a 234 cm (92") detachable, three-wire cord and plug. Calibrated Shakers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the temperature and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations. All units include a starter pack of 100 capillary tubes. Tubes are closed on one end.
Specifications for Product Family
Specifications
  • Catalog No:
  • 75840-104
  • 75840-108
  • Environmentally Preferable:
  • Waste Reducing
  • Waste Reducing
  • Capacity:
  • 3 capillary tubes
  • 3 capillary tubes
  • Operating conditions:
  • Unit can be run in conditions from 5 to 40 °C (41 to 104 °F), maximum 80% relative humidity, non-condensing
  • Unit can be run in conditions from 5 to 40 °C (41 to 104 °F), maximum 80% relative humidity, non-condensing
  • Temperature accuracy:
  • +/-0.5%
  • +/-0.5%
  • Temperature range:
  • 5° above ambient to 400 °C
  • 5° above ambient to 400 °C
  • Time:
  • mm/dd/yy hh:mm A/P (Real Time Clock)
  • mm/dd/yy hh:mm A/P (Real Time Clock)
  • Electrical:
  • 120V, 50/60Hz
  • 120V, 50/60Hz
  • Overall height:
  • 4.25"
  • 4.25"
  • Overall length:
  • 10.75"
  • 10.75"
  • Overall width:
  • 7.5"
  • 7.5"
  • Shipping weight:
  • 4 lbs.
  • 4 lbs.
  • Environmental certification:
  • RoHS
  • RoHS
Specifications
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Melting Point Apparatus
    Log in for availability
    Log in for availability
    Each
    Each - $2,281.11
    $2,281.11
    New Customer Promotional Price
    Each
       $2,281.11
    Specifications:
    Product available on GSA Advantage®
      Waste Reducing
    More
    Specifications
    Cat. No.
    75840-104
    Environmentally Preferable
    Waste Reducing
    Capacity
    3 capillary tubes
    Operating conditions
    Unit can be run in conditions from 5 to 40 °C (41 to 104 °F), maximum 80% relative humidity, non-condensing
    Temperature accuracy
    +/-0.5%
    Temperature range
    5° above ambient to 400 °C
    Time
    mm/dd/yy hh:mm A/P (Real Time Clock)
    Electrical
    120V, 50/60Hz
    Overall height
    4.25"
    Overall length
    10.75"
    Overall width
    7.5"
    Shipping weight
    4 lbs.
    Environmental certification
    RoHS
    Product available on GSA Advantage®
      Waste Reducing
  • Melting Point Apparatus with NIST Traceable Certificate
    Log in for availability
    Log in for availability
    Each
    Each - $4,772.39
    $4,772.39
    Each
       $4,772.39
    Specifications:
    Product available on GSA Advantage®
      Waste Reducing
    More
    Specifications
    Cat. No.
    75840-108
    Environmentally Preferable
    Waste Reducing
    Capacity
    3 capillary tubes
    Operating conditions
    Unit can be run in conditions from 5 to 40 °C (41 to 104 °F), maximum 80% relative humidity, non-condensing
    Temperature accuracy
    +/-0.5%
    Temperature range
    5° above ambient to 400 °C
    Time
    mm/dd/yy hh:mm A/P (Real Time Clock)
    Electrical
    120V, 50/60Hz
    Overall height
    4.25"
    Overall length
    10.75"
    Overall width
    7.5"
    Shipping weight
    4 lbs.
    Environmental certification
    RoHS
    Product available on GSA Advantage®
      Waste Reducing
Accessories
Product Information
  • Catalog #
    Details
    Availability
    Price
  • Capillary Tubes
    Capillary Tubes
    75840-018
    Capillary tubes with one end open for efficient sample collection; Tubes are 90mm long with an OD of 2mm and an ID of 1mm; Case of 2000, 90mm capillary tubes, open on one end
    Log in for availability
    Case of 2,000 - $510.10
Product Information
  • Catalog #
    Details
    Availability
    Price
  • Cooling Blocks
    Cooling Blocks
    75840-020
    Cooling blocks are designed to rapidly cool the oven back to room temperature; The copper material absorbs the heat and draws it out of the oven to provide efficient transition between tests involving various different melting points
    Log in for availability
    Each - $154.16